Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026257.3 | NP_080533.1 | 15 | 1482 | ||
GenBank mRNA | AK010492.1 | - | 8 | 959 | ||
AK015720.1 | - | 15 | 1466 | |||
AK048494.1 | - | 15 | 1484 | |||
BC019461.1 | - | 14 | 1338 | 4224033 |