Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026257.3 | NP_080533.1 | 12 | 1107 | ||
GenBank mRNA | AK010492.1 | - | 5 | 586 | ||
AK015720.1 | - | 12 | 1091 | |||
AK048494.1 | - | 12 | 1109 | |||
BC019461.1 | - | 11 | 963 | 4224033 |