Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163316.1 | NP_001156788.1 | 8 | 787 | ||
NM_026269.2 | NP_080545.2 | 8 | 978 | |||
GenBank mRNA | AK003551.1 | - | 8 | 927 | ||
BC019557.1 | - | 8 | 769 | 4163641 |