Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026272.3 | NP_080548.3 | 5 | 545 | ||
GenBank mRNA | AK013432.1 | - | 5 | 504 | ||
AK076232.1 | - | 5 | 528 | |||
AK081305.1 | - | 5 | 529 | |||
BC016090.1 | - | 5 | 484 | 4503713 | ||
BM951066.1 | - | 5 | 545 |