Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 5 | 1263 | ||
GenBank mRNA | AK032416.1 | - | 5 | 1226 | ||
AK076799.1 | - | 5 | 836 | |||
AK134544.1 | - | 2 | 130 | |||
AK141443.1 | - | 5 | 813 | |||
AK150941.1 | - | 4 | 738 | |||
AK151018.1 | - | 5 | 1233 | |||
AK152024.1 | - | 5 | 1233 | |||
AK152435.1 | - | 4 | 738 | |||
AK172567.1 | - | 5 | 1229 | |||
AK173313.1 | - | 5 | 882 | |||
AK212675.1 | - | 1 | 13 | |||
AK214813.1 | - | 1 | 13 | |||
BC010833.1 | - | 5 | 878 | 4235165 | ||
BC054121.1 | - | 5 | 1249 | 6393910 |