Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 14 | 2229 | ||
GenBank mRNA | AK013003.1 | - | 5 | 446 | ||
AK031153.1 | - | 5 | 545 | |||
AK032416.1 | - | 14 | 2192 | |||
AK076799.1 | - | 14 | 1802 | |||
AK141443.1 | - | 14 | 1779 | |||
AK150941.1 | - | 13 | 1704 | |||
AK151018.1 | - | 14 | 2199 | |||
AK152024.1 | - | 14 | 2199 | |||
AK152435.1 | - | 13 | 1704 | |||
AK172567.1 | - | 14 | 2195 | |||
AK173313.1 | - | 14 | 1848 | |||
BC010833.1 | - | 14 | 1844 | 4235165 | ||
BC054121.1 | - | 14 | 2215 | 6393910 |