Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163725.1 | NP_001157197.1 | 7 | 892 | ||
NM_026314.3 | NP_080590.3 | 9 | 1252 | |||
GenBank mRNA | AK005832.1 | - | 6 | 702 | ||
AK015345.1 | - | 5 | 431 | |||
AK153643.1 | - | 1 | 1215 | |||
AY428507.1 | - | 9 | 1252 | |||
AY464142.1 | - | 7 | 892 | |||
BC026462.1 | - | 9 | 1209 | 4483015 |