Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026319.3 | NP_080595.2 | 16 | 1457 | ||
GenBank mRNA | AK006492.1 | - | 7 | 564 | ||
BY706530.1 | - | 7 | 562 |