Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033135.3 | NP_001028307.2 | 3, 4 | 851 | ||
GenBank mRNA | AK155360.1 | - | 3, 4 | 910 | ||
AY155439.1 | - | 3, 4 | 443 | |||
BC115968.1 | - | 4, 5 | 431 | 40107454 | ||
CR759684.1 | - | 2, 3 | 229 |