Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026360.3 | NP_080636.2 | 9 | 939 | ||
GenBank mRNA | AK010310.1 | - | 9 | 932 | ||
AK076982.1 | - | 9 | 924 | |||
AK169595.1 | - | 7 | 562 | |||
BC006843.1 | - | 2 | 126 | |||
BC060168.1 | - | 9 | 919 | |||
BC095944.1 | - | 9 | 916 | 4935970 | ||
BC132242.1 | - | 9 | 931 | 40130519 | ||
BC132244.1 | - | 9 | 931 | 40130521 |