Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026360.3 | NP_080636.2 | 4, 5 | 466 | ||
GenBank mRNA | AK010310.1 | - | 4, 5 | 459 | ||
AK076982.1 | - | 4, 5 | 451 | |||
BC060168.1 | - | 4, 5 | 446 | |||
BC095944.1 | - | 4, 5 | 443 | 4935970 | ||
BC132242.1 | - | 4, 5 | 458 | 40130519 | ||
BC132244.1 | - | 4, 5 | 458 | 40130521 |