Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026374.3 | NP_080650.1 | 10 | 727 | ||
GenBank mRNA | AK012364.1 | - | 10 | 714 | ||
AK013858.1 | - | 10 | 773 | |||
AK078003.1 | - | 10 | 714 | |||
AK135449.1 | - | 10 | 802 | |||
AK135539.1 | - | 10 | 714 | |||
AK157129.1 | - | 10 | 782 | |||
AK212445.1 | - | 2 | 63 | |||
BC004033.1 | - | 2 | 94 | |||
BC024718.1 | - | 10 | 727 | 5012365 |