Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199146.1 | NP_001186075.1 | 3 | 504 | ||
NM_001199147.1 | NP_001186076.1 | 3 | 573 | |||
NM_153410.5 | NP_700459.2 | 3 | 504 | |||
GenBank mRNA | AK048691.1 | - | 3 | 498 | ||
AK082985.1 | - | 3 | 447 | |||
AK084631.1 | - | 3 | 446 | |||
AK190316.1 | - | 2 | 137 | |||
BC026486.1 | - | 3 | 564 | |||
BC027004.1 | - | 11 | ||||
BC071197.1 | - | 3 | 570 | 6465025 | ||
CA317518.1 | - | 3 | 522 |