Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026403.3 | NP_080679.3 | 10 | 1969 | ||
GenBank mRNA | AK011570.1 | - | 10 | 1959 | ||
AK015585.1 | - | 10 | 1719 | |||
AK032873.1 | - | 10 | 1953 | |||
AK037831.1 | - | 10 | 1957 | |||
AK050912.1 | - | 10 | 1960 | |||
AK083861.1 | - | 10 | 1838 | |||
AK139689.1 | - | 10 | 1922 | |||
AK146020.1 | - | 10 | 1924 | |||
AK146687.1 | - | 10 | 1941 | |||
AK152732.1 | - | 10 | 1943 | |||
AK154387.1 | - | 10 | 1961 | |||
AK159798.1 | - | 10 | 1943 | |||
BC018523.1 | - | 10 | 1949 | 3499986 |