Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024211.3 | NP_077173.1 | 8 | 1062 | ||
GenBank mRNA | AK003957.1 | - | 7 | 701 | ||
AK009487.1 | - | 8 | 1008 | |||
AK009824.1 | - | 8 | 962 | |||
AK168951.1 | - | 8 | 987 | |||
AK197579.1 | - | 1 | 75 | |||
AK199871.1 | - | 1 | 75 | |||
AK202008.1 | - | 1 | 75 | |||
AK213339.1 | - | 1 | 75 | |||
BC003455.1 | - | 8 | 1038 | 2648336 | ||
BC019631.1 | - | 8 | 1016 | 4023179 |