Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167829.1 | NP_001161301.1 | 8 | 843 | ||
NM_026437.3 | NP_080713.2 | 8 | 864 | |||
GenBank mRNA | AK004643.1 | - | 7 | 784 | ||
AK007867.1 | - | 8 | 824 | |||
AK145769.1 | - | 8 | 830 | |||
AK168474.1 | - | 7 | 778 | |||
BC013514.1 | - | 8 | 823 | 4235293 |