Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026447.4 | NP_080723.3 | 3 | 407 | ||
NM_198931.3 | NP_945149.2 | 3 | 411 | |||
GenBank mRNA | AB474373.1 | - | 3 | 446 | ||
AK013149.1 | - | 3 | 340 | |||
AK046387.1 | - | 306 | ||||
AK087999.1 | - | 3 | 445 | |||
AK186072.1 | - | 2 | 79 | |||
AK217232.1 | - | 2 | 79 | |||
AY332616.1 | - | 3 | 414 | |||
BC058248.1 | - | 3 | 333 |