Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164139.1 | NP_001157611.1 | 3 | 1355 | ||
NM_001164140.1 | NP_001157612.1 | 2 | 1274 | |||
NM_001164141.1 | NP_001157613.1 | 3 | 1363 | |||
NM_026457.2 | NP_080733.2 | 2 | 1213 | |||
GenBank mRNA | AK005703.1 | - | 2 | 1211 | ||
AK006329.1 | - | 1 | 943 | |||
AK007022.1 | - | 2 | 1192 | |||
AY092416.1 | - | 2 | 1347 | |||
BC109147.2 | - | 2 | 1192 | 40045644 |