Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026458.4 | NP_080734.3 | 3 | 346 | ||
GenBank mRNA | AK007156.1 | - | 3 | 307 | ||
AK015996.1 | - | 3 | 303 | |||
AK016294.1 | - | 3 | 301 | |||
AK029665.1 | - | 3 | 350 | |||
AY243470.1 | - | 3 | 346 | |||
BC116805.1 | - | 3 | 277 | |||
BC116809.1 | - | 3 | 277 |