Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175127.2 | NP_780336.1 | 3, 4 | 534 | ||
GenBank mRNA | AK076211.1 | - | 3, 4 | 540 | ||
AK077653.1 | - | 3, 4 | 547 | |||
AK122295.1 | - | 3, 4 | 519 | |||
AK162091.1 | - | 3, 4 | 503 | |||
AK162188.1 | - | 3, 4 | 415 | |||
BC055458.1 | - | 2, 3 | 236 | |||
BC094542.1 | - | 3, 4 | 534 | 6408744 |