Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nifk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026472.4 | NP_080748.3 | 7 | 827 | ||
GenBank mRNA | AB056870.1 | - | 7 | 777 | ||
AK009440.1 | - | 7 | 794 | |||
AK077770.1 | - | 7 | 822 | |||
AK081486.1 | - | 7 | 794 | |||
AY030275.1 | - | 7 | 798 | |||
BC058559.1 | - | 7 | 806 | 6314031 |