Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 8, 9 | 1605 | ||
GenBank mRNA | AK008785.1 | - | 5, 6 | 535 | ||
AK013337.1 | - | 8, 9 | 1593 | |||
BC018177.1 | - | 8, 9 | 1599 | |||
BC049270.1 | - | 8, 9 | 1581 | 3988645 |