Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037098.1 | NP_001032175.1 | 2 | 1062 | ||
NM_026495.3 | NP_080771.3 | 2 | 539 | |||
GenBank mRNA | AK002651.1 | - | 2 | 513 | ||
AK043047.1 | - | 2 | 487 | |||
AK147293.1 | - | 2 | 539 | |||
AK147310.1 | - | 2 | 643 | |||
AK157946.1 | - | 2 | 646 | |||
AK189806.1 | - | 1 | 79 | |||
AK202844.1 | - | 1 | 45 | |||
AK219019.1 | - | 1 | 108 | |||
BC022103.1 | - | 2 | 521 | 5067063 | ||
BC070453.1 | - | 2 | 843 |