Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026498.2 | NP_080774.2 | 5 | 660 | ||
GenBank mRNA | AB049945.1 | - | 5 | 384 | ||
AK002993.1 | - | 5 | 497 | |||
AK152027.1 | - | 5 | 451 | |||
AK160207.1 | - | 5 | 440 | |||
AK166241.1 | - | 5 | 448 | |||
BC005701.1 | - | 5 | 402 | |||
BC028279.1 | - | 5 | 452 | 5370270 |