Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080556.3 | NP_542123.3 | 15 | 1902 | ||
GenBank mRNA | AK010325.1 | - | 15 | 1859 | ||
AK030340.1 | - | 15 | 1833 | |||
AK048012.1 | - | 15 | 1821 | |||
AK050338.1 | - | 15 | 1849 | |||
AK075720.1 | - | 15 | 1902 | |||
AK088905.1 | - | 15 | 1874 | |||
AK159757.1 | - | 15 | 1847 | |||
BC003862.1 | - | 15 | 1841 | 3488326 |