Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114096.1 | NP_001107568.1 | 25 | 2748 | ||
NM_001114097.1 | NP_001107569.1 | 24 | 2655 | |||
NM_198160.2 | NP_937803.1 | 24 | 2655 | |||
GenBank mRNA | AK089461.1 | - | 24 | 2590 | ||
AK146971.1 | - | 25 | 2677 | |||
AK170530.1 | - | 24 | 2589 | |||
BC040363.1 | - | 24 | 2570 | |||
BC058720.1 | - | 24 | 2597 | 5705438 | ||
BC062102.1 | - | 13 | 1454 |