Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114096.1 | NP_001107568.1 | 9, 10 | 933 | ||
NM_001114097.1 | NP_001107569.1 | 9, 10 | 933 | |||
NM_198160.2 | NP_937803.1 | 9, 10 | 933 | |||
GenBank mRNA | AK146971.1 | - | 9, 10 | 862 | ||
AK215230.1 | - | 1, 2 | 41 | |||
BC040363.1 | - | 9, 10 | 848 | |||
BC058720.1 | - | 9, 10 | 875 | 5705438 |