Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026576.3 | NP_080852.2 | 6 | 2689 | ||
GenBank mRNA | AK018594.1 | - | 2 | 1986 | ||
AK034138.1 | - | 6 | 2682 | |||
AK160833.1 | - | 1 | 2728 | |||
BC028916.1 | - | 6 | 2684 | 3676092 |