Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172399.3 | NP_765987.2 | 4 | 1621 | ||
GenBank mRNA | AK029210.1 | - | 4 | 1622 | ||
AK049802.1 | - | 4 | 1646 | |||
BC047154.1 | - | 4 | 1595 | 6489681 | ||
CK618056.1 | - | 1 | 167 |