Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026620.3 | NP_080896.2 | 3, 4 | 370 | ||
GenBank mRNA | AK019190.1 | - | 3, 4 | 371 | ||
AK019253.1 | - | 3, 4 | 371 | |||
AK039305.1 | - | 3, 4 | 1900 | |||
BC048087.1 | - | 3, 4 | 360 | 6439798 |