Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026642.2 | NP_080918.2 | 1 | 1196 | ||
GenBank mRNA | AK019481.1 | - | 1 | 1172 | ||
AK031429.1 | - | 1 | 1148 | |||
AK076857.1 | - | 1 | 1166 | |||
AK079291.1 | - | 1 | 1154 | |||
BC052533.1 | - | 1 | 1166 | 5255779 | ||
BC055850.1 | - | 1 | 1154 | 4193356 |