Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290658.1 | NP_001277587.1 | 18 | 1707 | ||
NM_026651.2 | NP_080927.1 | 19 | 1793 | |||
NM_029786.2 | NP_084062.2 | 18 | 1694 | |||
GenBank mRNA | AB053220.1 | - | 19 | 1775 | ||
AK002638.1 | - | 18 | 1686 | |||
AK015478.1 | - | 18 | 1709 | |||
AK019640.1 | - | 18 | 1699 | |||
AK178221.1 | - | 3 | 157 | |||
AK180366.1 | - | 3 | 157 | |||
AK210391.1 | - | 3 | 157 | |||
AK212205.1 | - | 3 | 157 | |||
BC011201.1 | - | 19 | 1716 | 4190368 | ||
BC023265.1 | - | 16 | 1381 |