Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310057M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026655.3 | NP_080931.1 | 4 | 647 | ||
GenBank mRNA | AK019097.2 | - | 4 | 649 | ||
AK088850.1 | - | 4 | 683 | |||
AK148008.1 | - | 656 | ||||
BC044749.2 | - | 4 | 655 | 5102741 | ||
BC052432.1 | - | 4 | 645 |