Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 3 | 1051 | ||
GenBank mRNA | AK012153.1 | - | 3 | 720 | ||
AK018758.2 | - | 3 | 761 | |||
AK145674.1 | - | 3 | 692 | |||
AK148065.1 | - | 3 | 748 | |||
AK152899.1 | - | 3 | 1099 | |||
AK154979.1 | - | 3 | 740 | |||
AK170079.1 | - | 3 | 763 | |||
AX799084.1 | - | 3 | 762 | |||
BC052433.1 | - | 3 | 1051 | 6837097 |