Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 12 | 2210 | ||
GenBank mRNA | AK012153.1 | - | 12 | 1880 | ||
AK018758.2 | - | 12 | 1920 | |||
AK145674.1 | - | 12 | 1851 | |||
AK148065.1 | - | 12 | 1909 | |||
AK152899.1 | - | 12 | 2258 | |||
AK154979.1 | - | 12 | 1899 | |||
AK170079.1 | - | 12 | 1922 | |||
AK205163.1 | - | 1 | 57 | |||
AX799084.1 | - | 12 | 1921 | |||
BC003206.1 | - | 7 | 922 | |||
BC013054.1 | - | 2 | 328 | |||
BC052433.1 | - | 12 | 2210 | 6837097 |