Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146171.1 | NP_666283.1 | 15 | 2017 | ||
GenBank mRNA | AK019259.2 | - | 1 | 181 | ||
AK129068.1 | - | 15 | 1982 | |||
AK156143.1 | - | 15 | 2048 | |||
AK213703.1 | - | 1 | 106 | |||
BC006673.1 | - | 8 | 1163 | |||
BC008592.1 | - | 6 | 911 | |||
BC023750.1 | - | 1 | 46 | |||
BC025460.1 | - | 15 | 2037 | |||
BC026982.1 | - | 15 | 2017 | 5364933 | ||
BC029133.1 | - | 15 | 1974 | 4951596 |