Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164742.1 | NP_001158214.1 | 2 | 428 | ||
NM_026687.2 | NP_080963.2 | 3 | 568 | |||
GenBank mRNA | AB233454.1 | - | 3 | 568 | ||
AK002574.1 | - | 2 | 456 | |||
AK165409.1 | - | 2 | 406 | |||
BC119526.1 | - | 2 | 396 | 40056096 |