Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026713.3 | NP_080989.2 | 5 | 744 | ||
GenBank mRNA | AF384162.1 | - | 4 | 621 | ||
AK002693.1 | - | 4 | 693 | |||
BC031920.1 | - | 768 | ||||
BC106135.1 | - | 4 | 692 | 5041577 | ||
BE626067.1 | - | 3 | 471 |