Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026744.3 | NP_081020.1 | 2, 3 | 217 | ||
GenBank mRNA | AK003549.1 | - | 2, 3 | 218 | ||
BC022162.1 | - | 2, 3 | 242 | 5148541 | ||
BC065995.1 | - | 1440 | 5688315 |