Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043335.1 | NP_001036800.1 | 2 | 524 | ||
NM_001043336.2 | NP_001036801.1 | 2 | 141 | |||
NM_001286346.1 | NP_001273275.1 | 2 | 278 | |||
NM_001286347.1 | NP_001273276.1 | 2 | 141 | |||
GenBank mRNA | AK044662.1 | - | 2 | 263 | ||
AK054227.1 | - | 2 | 474 | |||
BC053094.1 | - | 2 | 358 | 6400458 | ||
BC059839.1 | - | 2 | 119 | 6417403 | ||
BC079582.1 | - | 2 | 177 | 5702743 | ||
BC125289.1 | - | 2 | 112 | 40129804 | ||
BC144757.1 | - | 2 | 112 | |||
BC150975.1 | - | 2 | 278 | 9087888 | ||
BY124470.1 | - | 2 | 142 | |||
CD350405.1 | - | 2 | 524 |