Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023479.2 | NP_075968.2 | 17 | 1596 | ||
GenBank mRNA | AF308696.2 | - | 17 | 1534 | ||
AK004136.1 | - | 5 | 332 | |||
AK156730.1 | - | 17 | 1597 | |||
AK172604.1 | - | 15 | 2044 | |||
BC048235.1 | - | 17 | 1530 | 6493124 |