Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026795.3 | NP_081071.1 | 2 | 316 | ||
NM_178056.3 | NP_835157.1 | 3 | 409 | |||
GenBank mRNA | AK003917.1 | - | 2 | 275 | ||
AK077858.1 | - | 1023 | ||||
BC024620.1 | - | 2 | 269 | 4192712 |