Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138667.3 | NP_619608.1 | 2 | 424 | ||
GenBank mRNA | AK086230.1 | - | 1 | 1050 | ||
AK089164.1 | - | 2 | 424 | |||
AY093701.1 | - | 1 | 84 | |||
BC004813.1 | - | 1 | 91 | 3586132 | ||
CK391470.1 | - | 3 | 290 |