Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 8 | 797 | ||
GenBank mRNA | AK003990.1 | - | 8 | 753 | ||
AK039949.1 | - | 8 | 797 | |||
AK041783.1 | - | 791 | ||||
AK084395.1 | - | 8 | 791 | |||
AK086633.1 | - | 791 | ||||
AK144975.1 | - | 8 | 798 | |||
AK159740.1 | - | 8 | 809 | |||
AK161378.1 | - | 8 | 787 | |||
AK168510.1 | - | 8 | 753 | |||
AK169066.1 | - | 8 | 787 | |||
AK169351.1 | - | 8 | 754 | |||
AK177148.1 | - | 5 | 427 | |||
AK179856.1 | - | 5 | 384 | |||
AK181035.1 | - | 5 | 410 | |||
AK181279.1 | - | 5 | 395 | |||
BC119178.1 | - | 8 | 791 | 8733927 | ||
BC119180.1 | - | 8 | 791 | 8733929 |