Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 22 | 2175 | ||
NM_152808.3 | NP_690021.1 | 22 | 2177 | |||
GenBank mRNA | AK041533.1 | - | 22 | 2134 | ||
AK048648.1 | - | 22 | 2162 | |||
AK153992.1 | - | 22 | 2148 | |||
AK166695.1 | - | 22 | 2178 | |||
BC013213.1 | - | 15 | 1570 | |||
BC023763.1 | - | 16 | 1412 | |||
BC031535.1 | - | 22 | 2153 | 3991609 |