Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197987.2 | NP_932104.1 | 13 | 1546 | ||
GenBank mRNA | AB093271.1 | - | 13 | 1517 | ||
AK140822.1 | - | 13 | 1547 | |||
BC058678.1 | - | 13 | 1544 | 6834513 | ||
BC059070.1 | - | 13 | 1512 | 6825999 | ||
BC078416.1 | - | 13 | 1531 |