Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144524.2 | NP_653107.2 | 3 | 797 | ||
GenBank mRNA | AK143549.1 | - | 3 | 798 | ||
BC019148.1 | - | 3 | 812 | 5052974 |