Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028390.3 | NP_082666.2 | 4 | 904 | ||
GenBank mRNA | BC027141.1 | - | 4 | 868 | ||
BC032164.1 | - | 4 | 922 | 5346703 |