Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175132.3 | NP_780341.1 | 4 | 2764 | ||
GenBank mRNA | AK053054.1 | - | 4 | 2764 | ||
AK084541.1 | - | 2 | 2051 | |||
BC137972.1 | - | 4 | 2735 | 8860991 | ||
BC145070.1 | - | 4 | 2734 |