Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026836.2 | NP_081112.1 | 3 | 420 | ||
GenBank mRNA | AK004168.1 | - | 2 | 67 | ||
AK050689.1 | - | 3 | 421 | |||
AK133851.1 | - | 4 | 1404 | |||
BC005603.1 | - | 3 | 399 | 3483315 |